Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021643.3 | NP_067675.1 | 1 | 1657 | ||
GenBank mRNA | AK313237.1 | - | 1 | 707 | ||
AY245544.1 | - | 1 | 1640 | |||
BC002637.2 | - | 1 | 707 | 3607549 | ||
D87119.1 | - | 1 | 1446 |