Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MELK SIRNA Assays ›
Gene Symbol:
MELK
Gene Name:
maternal embryonic leucine zipper kinase
Gene Aliases:
HPK38
Chromosome Location:
Chr.9: 36572862 - 36677683 on Build GRCh38
UniGene ID:
Hs.184339
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
103433
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MELK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256685.1
NP_001243614.1
7
675
NM_001256687.1
NP_001243616.1
6
531
NM_001256688.1
NP_001243617.1
5
462
NM_001256689.1
NP_001243618.1
6
589
NM_001256690.1
NP_001243619.1
5
472
NM_001256691.1
NP_001243620.1
5
445
NM_001256692.1
NP_001243621.1
4
349
NM_001256693.1
NP_001243622.1
5
493
NM_014791.3
NP_055606.1
7
675
NR_046337.1
6
589
XM_011518076.2
XP_011516378.1
7
854
XM_011518077.1
XP_011516379.1
8
750
XM_011518078.2
XP_011516380.1
8
937
XM_011518079.1
XP_011516381.1
7
627
XM_011518080.1
XP_011516382.1
7
642
XM_011518081.2
XP_011516383.1
7
660
XM_011518082.2
XP_011516384.1
6
768
XM_011518083.2
XP_011516385.1
7
851
XM_011518084.2
XP_011516386.1
6
463
XM_011518085.1
XP_011516387.1
5
459
XM_011518086.2
XP_011516388.1
7
642
GenBank mRNA
AB183427.1
-
6
540
AB183428.1
-
5
425
AK222811.1
-
7
674
AK293284.1
-
5
427
AK293423.1
-
5
410
AK293447.1
-
5
429
AK296820.1
-
6
588
AK299164.1
-
7
627
AK300761.1
-
6
496
AK300821.1
-
6
543
AK301131.1
-
5
425
AK302374.1
-
4
316
AK309470.1
-
5
450
BC014039.2
-
7
629
4547136
D79997.1
-
7
661
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
apoptotic process
cell proliferation
intrinsic apoptotic signaling pathway in response to oxidative stress
peptidyl-tyrosine phosphorylation
hemopoiesis
intracellular signal transduction
positive regulation of apoptotic process
protein autophosphorylation
neural precursor cell proliferation
protein serine/threonine kinase activity
non-membrane spanning protein tyrosine kinase activity
calcium ion binding
protein binding
ATP binding
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9xzrh:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline