Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260504.1 | NP_001247433.1 | 7 | 1069 | ||
NM_001260505.1 | NP_001247434.1 | 7 | 793 | |||
NM_031414.4 | NP_113602.2 | 7 | 793 | |||
NM_032944.3 | NP_116562.2 | 7 | 1037 | |||
NR_048542.1 | 7 | 785 | ||||
XM_011515449.2 | XP_011513751.2 | 7 | 719 | |||
XM_011515450.1 | XP_011513752.1 | 7 | 1040 | |||
GenBank mRNA | AF285599.1 | - | 7 | 689 | ||
AF332194.1 | - | 7 | 1072 | |||
AK057271.1 | - | 7 | 741 | |||
AK292474.1 | - | 7 | 685 | |||
AK302689.1 | - | 7 | 685 | |||
BC033018.1 | - | 7 | 687 | |||
BC059374.1 | - | 7 | 700 | 5268051 |