Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000459.4 | NP_000450.2 | 13 | 2483 | ||
NM_001290077.1 | NP_001277006.1 | 12 | 2354 | |||
NM_001290078.1 | NP_001277007.1 | 11 | 2042 | |||
XM_005251561.1 | XP_005251618.1 | 13 | 2150 | |||
XM_005251563.1 | XP_005251620.1 | 12 | 2021 | |||
GenBank mRNA | AB086825.1 | - | 12 | 2010 | ||
AB208796.1 | - | 13 | 2161 | |||
AK291775.1 | - | 13 | 2171 | |||
AK294887.1 | - | 11 | 2043 | |||
AK295043.1 | - | 12 | 2080 | |||
BC035514.1 | - | 13 | 2158 | 5228999 | ||
H25908.1 | - | 1 | 123 | |||
L06139.1 | - | 13 | 2181 |