Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204.6 | NP_001195.2 | 7, 8 | 2120 | ||
XM_011511687.1 | XP_011509989.1 | 7, 8 | 2137 | |||
GenBank mRNA | AK292430.1 | - | 7, 8 | 1384 | ||
BC052985.2 | - | 7, 8 | 1546 | 6149698 | ||
D50516.1 | - | 7, 8 | 1380 | |||
U20165.1 | - | 7, 8 | 1008 | |||
Z48923.1 | - | 7, 8 | 1353 |