Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018638.4 | NP_061108.2 | 6 | 1203 | ||
XM_017019580.1 | XP_016875069.1 | 6 | 1200 | |||
GenBank mRNA | AF207600.2 | - | 6 | 1269 | ||
AK074425.1 | - | 6 | 1148 | |||
AL137750.1 | - | 6 | 1152 | |||
BC066907.1 | - | 6 | 1203 | 5260109 |