Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021643.3 | NP_067675.1 | 3 | 2344 | ||
NR_027303.1 | 3 | 777 | ||||
GenBank mRNA | AK299146.1 | - | 3 | 751 | ||
AK313237.1 | - | 3 | 1394 | |||
AY245544.1 | - | 3 | 2327 | |||
BC002637.2 | - | 3 | 1394 | 3607549 | ||
D87119.1 | - | 3 | 2133 |