Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304349.1 | NP_001291278.1 | 4, 5 | 978 | ||
NM_002730.3 | NP_002721.1 | 4, 5 | 544 | |||
NM_207518.2 | NP_997401.1 | 4, 5 | 353 | |||
XM_017026948.1 | XP_016882437.1 | 4, 5 | 400 | |||
GenBank mRNA | AF208004.1 | - | 4, 5 | 338 | ||
AF239744.1 | - | 4, 5 | 483 | |||
AK131561.1 | - | 4, 5 | 978 | |||
AK290147.1 | - | 5, 6 | 426 | |||
AK292284.1 | - | 4, 5 | 353 | |||
AK301264.1 | - | 4, 5 | 713 | |||
BC039846.1 | - | 4, 5 | 540 | 6066764 | ||
BC108259.1 | - | 4, 5 | 541 | 6144307 | ||
BU543573.1 | - | 544 | ||||
M80335.1 | - | 3, 4 | 293 | |||
X07767.1 | - | 4, 5 | 424 |