Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006944.1 | NP_001006945.1 | 8 | 928 | ||
NM_001300802.1 | NP_001287731.1 | 8 | 928 | |||
NM_001318361.1 | NP_001305290.1 | 8 | 882 | |||
NM_003942.2 | NP_003933.1 | 8 | 928 | |||
XM_005274380.1 | XP_005274437.1 | 8 | 908 | |||
XM_017018527.1 | XP_016874016.1 | 4 | 328 | |||
GenBank mRNA | AF074715.1 | - | 7 | 797 | ||
AJ010119.1 | - | 8 | 910 | |||
AK223561.1 | - | 8 | 933 | |||
AK292163.1 | - | 8 | 909 | |||
AK298507.1 | - | 8 | 853 | |||
AK313567.1 | - | 8 | 908 | |||
BC020451.1 | - | 5 | 483 | |||
BC028079.1 | - | 8 | 892 | |||
BC047896.1 | - | 8 | 907 | 5763859 |