Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMPR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001203.2 | NP_001194.1 | 8 | 777 | ||
NM_001256792.1 | NP_001243721.1 | 6 | 673 | |||
NM_001256793.1 | NP_001243722.1 | 6 | 614 | |||
NM_001256794.1 | NP_001243723.1 | 6 | 657 | |||
XM_011532201.2 | XP_011530503.1 | 7 | 656 | |||
XM_017008558.1 | XP_016864047.1 | 8 | 848 | |||
XM_017008559.1 | XP_016864048.1 | 9 | 838 | |||
XM_017008560.1 | XP_016864049.1 | 6 | 730 | |||
XM_017008561.1 | XP_016864050.1 | 7 | 687 | |||
GenBank mRNA | AK293008.1 | - | 6 | 657 | ||
AK299930.1 | - | 6 | 614 | |||
AK313642.1 | - | 6 | 673 | |||
BC047773.1 | - | 5 | 573 | 6202951 | ||
BC069796.1 | - | 7 | 689 | 7262484 | ||
BC069803.1 | - | 7 | 689 | 7262496 | ||
D89675.1 | - | 5 | 521 | |||
U89326.1 | - | 8 | 776 |