Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301061.1 | NP_001287990.1 | 4, 5 | 551 | ||
NM_001707.3 | NP_001698.2 | 3, 4 | 429 | |||
NR_036682.1 | 3, 4 | 412 | ||||
GenBank mRNA | AJ223979.1 | - | 3, 4 | 308 | ||
AK290091.1 | - | 3, 4 | 337 | |||
AL525884.3 | - | 4, 5 | 492 | |||
AL553260.3 | - | 3, 4 | 696 | |||
BC000956.2 | - | 3, 4 | 388 | 3450946 | ||
BC001967.2 | - | 3, 4 | 295 | 3461829 | ||
BC009548.1 | - | 3, 4 | 339 | 3897805 | ||
BI603803.1 | - | 3, 4 | 425 | |||
BQ109412.1 | - | 3, 4 | 361 | |||
DA688517.1 | - | 3, 4 | 412 | |||
X89985.1 | - | 3, 4 | 309 |