Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003821.5 | NP_003812.1 | 4 | 936 | ||
XM_005251092.3 | XP_005251149.1 | 3 | 746 | |||
XM_011517357.2 | XP_011515659.1 | 2 | 269 | |||
GenBank mRNA | AF027706.1 | - | 4 | 846 | ||
AF064824.1 | - | 4 | 864 | |||
AF078530.1 | - | 4 | 622 | |||
AK075213.1 | - | 4 | 840 | |||
AK301448.1 | - | 3 | 695 | |||
AY358813.1 | - | 3 | 693 | |||
AY358814.1 | - | 4 | 873 | |||
AY524045.1 | - | 4 | 709 | |||
BC004553.2 | - | 4 | 824 | 4026156 | ||
KU178437.1 | - | 4 | 622 | |||
KU178438.1 | - | 3 | 468 |