Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006944.1 | NP_001006945.1 | 9, 10 | 1162 | ||
NM_001300802.1 | NP_001287731.1 | 9, 10 | 1162 | |||
NM_001318361.1 | NP_001305290.1 | 9, 10 | 1116 | |||
NM_003942.2 | NP_003933.1 | 9, 10 | 1162 | |||
XM_005274380.1 | XP_005274437.1 | 8, 9 | 977 | |||
XM_017018527.1 | XP_016874016.1 | 5, 6 | 562 | |||
GenBank mRNA | AF074715.1 | - | 8, 9 | 1031 | ||
AJ010119.1 | - | 9, 10 | 1144 | |||
AK223561.1 | - | 9, 10 | 1167 | |||
AK292163.1 | - | 9, 10 | 1143 | |||
AK298507.1 | - | 9, 10 | 1087 | |||
AK313567.1 | - | 9, 10 | 1142 | |||
BC020451.1 | - | 6, 7 | 717 | |||
BC028079.1 | - | 9, 10 | 1126 | |||
BC047896.1 | - | 9, 10 | 1141 | 5763859 |