Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001711.5 | NP_001702.1 | 8 | 1668 | ||
XM_017029724.1 | XP_016885213.1 | 7 | 1447 | |||
GenBank mRNA | AK092954.1 | - | 8 | 1447 | ||
AK093023.1 | - | 8 | 1502 | |||
AK094059.1 | - | 9 | 1477 | |||
AK223302.1 | - | 8 | 1548 | |||
BC002416.2 | - | 8 | 1561 | 3162633 | ||
BC004244.1 | - | 8 | 1571 | 3503374 | ||
U11686.1 | - | 1 | 353 |