Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143676.1 | NP_001137148.1 | 6, 7 | 1224 | ||
NM_001143677.1 | NP_001137149.1 | 4, 5 | 475 | |||
NM_001143678.1 | NP_001137150.1 | 4, 5 | 654 | |||
NM_001291995.1 | NP_001278924.1 | 4, 5 | 430 | |||
NM_005627.3 | NP_005618.2 | 4, 5 | 430 | |||
XM_011536071.1 | XP_011534373.1 | 7, 8 | 1006 | |||
GenBank mRNA | AF153609.1 | - | 4, 5 | 383 | ||
AK055077.1 | - | 6, 7 | 1224 | |||
AK098509.1 | - | 4, 5 | 398 | |||
AK295422.1 | - | 4, 5 | 398 | |||
AK298688.1 | - | 4, 5 | 713 | |||
BC001263.1 | - | 4, 5 | 383 | 3459056 | ||
DB450096.1 | - | 4, 5 | 429 | |||
EU518415.1 | - | 6, 7 | 626 | |||
FM205707.1 | - | 4, 5 | 418 | |||
FM205708.1 | - | 4, 5 | 453 | |||
FM205709.1 | - | 6, 7 | 1073 | |||
FM205710.1 | - | 3, 4 | 542 | |||
Y10032.1 | - | 4, 5 | 383 |