Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256312.1 | NP_001243241.1 | 7 | 889 | ||
NM_006281.3 | NP_006272.2 | 5 | 546 | |||
XM_011517248.2 | XP_011515550.2 | 6 | 2519 | |||
XM_011517251.1 | XP_011515553.1 | 5 | 560 | |||
XM_011517253.2 | XP_011515555.1 | 3 | 229 | |||
XM_011517257.2 | XP_011515559.2 | 6 | 2519 | |||
XM_017013756.1 | XP_016869245.1 | 6 | 2519 | |||
XM_017013757.1 | XP_016869246.1 | 6 | 2519 | |||
XM_017013758.1 | XP_016869247.1 | 5 | 560 | |||
XM_017013759.1 | XP_016869248.1 | 7 | 895 | |||
XM_017013760.1 | XP_016869249.1 | 6 | 758 | |||
XM_017013761.1 | XP_016869250.1 | 5 | 560 | |||
XM_017013762.1 | XP_016869251.1 | 6 | 2519 | |||
GenBank mRNA | AK075229.1 | - | 7 | 846 | ||
AK131363.1 | - | 7 | 889 | |||
AK291837.1 | - | 5 | 510 | |||
AK312660.1 | - | 5 | 539 | |||
BC010640.2 | - | 5 | 535 | 3868822 | ||
U26424.1 | - | 5 | 543 | |||
U60206.1 | - | 5 | 480 | |||
Z25422.1 | - | 2 | 120 |