Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001713.2 | NP_001704.2 | 4 | 404 | ||
GenBank mRNA | AK315224.1 | - | 4 | 376 | ||
BC012616.1 | - | 4 | 380 | 4045543 | ||
DA370221.1 | - | 4 | 405 | |||
EU794592.1 | - | 4 | 404 | |||
U50929.1 | - | 4 | 325 |