Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261.3 | NP_001252.1 | 7 | 953 | ||
XM_017014184.1 | XP_016869673.1 | 7 | 1013 | |||
GenBank mRNA | AB451289.1 | - | 7 | 830 | ||
AB451421.1 | - | 7 | 830 | |||
AK313828.1 | - | 7 | 882 | |||
BC001968.1 | - | 7 | 890 | 3460767 | ||
BT019903.1 | - | 7 | 830 | |||
L25676.1 | - | 7 | 894 | |||
X80230.1 | - | 7 | 928 |