Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006944.1 | NP_001006945.1 | 11 | 1330 | ||
NM_001300802.1 | NP_001287731.1 | 11 | 1327 | |||
NM_001318361.1 | NP_001305290.1 | 11 | 1302 | |||
NM_003942.2 | NP_003933.1 | 11 | 1348 | |||
XM_005274380.1 | XP_005274437.1 | 10 | 1163 | |||
XM_017018527.1 | XP_016874016.1 | 7 | 748 | |||
GenBank mRNA | AF074715.1 | - | 10 | 1199 | ||
AJ010119.1 | - | 11 | 1330 | |||
AK223561.1 | - | 11 | 1332 | |||
AK292163.1 | - | 11 | 1329 | |||
AK298507.1 | - | 11 | 1273 | |||
AK313567.1 | - | 11 | 1307 | |||
BC020451.1 | - | 8 | 903 | |||
BC028079.1 | - | 11 | 1312 |