Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005443.4 | NP_005434.4 | 4, 5 | 630 | ||
XM_011532400.1 | XP_011530702.1 | 4, 5 | 610 | |||
XM_011532401.1 | XP_011530703.1 | 4, 5 | 696 | |||
GenBank mRNA | AF016496.1 | - | 4, 5 | 546 | ||
AF033026.1 | - | 4, 5 | 572 | |||
AF105227.1 | - | 4, 5 | 600 | |||
AK292774.1 | - | 4, 5 | 580 | |||
AK297981.1 | - | 3, 5 | 453 | |||
AU132232.1 | - | 4, 5 | 630 | |||
BC011392.1 | - | 4, 5 | 503 | 3869484 | ||
BC032513.1 | - | 4, 5 | 599 | |||
BC040940.1 | - | 4, 5 | 558 | |||
BC050627.1 | - | 4, 5 | 566 | 5556165 | ||
BQ437924.1 | - | 4, 5 | 558 | |||
CR457028.1 | - | 4, 5 | 546 | |||
Y10387.1 | - | 4, 5 | 582 |