Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256312.1 | NP_001243241.1 | 8, 9 | 1175 | ||
NM_006281.3 | NP_006272.2 | 6, 7 | 832 | |||
XM_011517248.2 | XP_011515550.2 | 7, 8 | 2805 | |||
XM_011517251.1 | XP_011515553.1 | 6, 7 | 846 | |||
XM_011517253.2 | XP_011515555.1 | 4, 5 | 515 | |||
XM_017013756.1 | XP_016869245.1 | 7, 8 | 2805 | |||
XM_017013757.1 | XP_016869246.1 | 7, 8 | 2805 | |||
XM_017013758.1 | XP_016869247.1 | 6, 7 | 846 | |||
XM_017013759.1 | XP_016869248.1 | 8, 9 | 1181 | |||
XM_017013760.1 | XP_016869249.1 | 7, 8 | 1044 | |||
GenBank mRNA | AK131363.1 | - | 8, 9 | 1175 | ||
AK291837.1 | - | 6, 7 | 796 | |||
AK312660.1 | - | 6, 7 | 825 | |||
BC010640.2 | - | 6, 7 | 821 | 3868822 | ||
U26424.1 | - | 6, 7 | 829 | |||
U60206.1 | - | 6, 7 | 766 |