Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDADC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193478.1 | NP_001180407.1 | 8 | 1528 | ||
NM_030911.3 | NP_112173.1 | 8 | 1528 | |||
NR_036439.1 | 7 | 1350 | ||||
XM_006719871.3 | XP_006719934.1 | 7 | 1297 | |||
XM_011535250.2 | XP_011533552.1 | 8 | 1492 | |||
XM_011535251.1 | XP_011533553.1 | 8 | 1472 | |||
XM_011535253.1 | XP_011533555.1 | 8 | 1424 | |||
XM_017020778.1 | XP_016876267.1 | 7 | 1231 | |||
XM_017020779.1 | XP_016876268.1 | 9 | 1648 | |||
XM_017020780.1 | XP_016876269.1 | 9 | 1995 | |||
XM_017020781.1 | XP_016876270.1 | 8 | 1467 | |||
XM_017020782.1 | XP_016876271.1 | 9 | 1571 | |||
XM_017020783.1 | XP_016876272.1 | 7 | 1692 | |||
XM_017020784.1 | XP_016876273.1 | 6 | 1263 | |||
GenBank mRNA | AK297400.1 | - | 7 | 1314 | ||
AK312835.1 | - | 8 | 1498 | |||
AL137554.1 | - | 2 | 204 | |||
AY027525.1 | - | 8 | 1477 | |||
BC048092.1 | - | 9 | 1496 | |||
BC125202.1 | - | 8 | 1442 | 40123430 |