Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206999.1 | NP_001193928.1 | 2 | 141 | ||
NM_007174.2 | NP_009105.1 | 2 | 141 | |||
XM_006719206.2 | XP_006719269.1 | 2 | 168 | |||
XM_011537783.1 | XP_011536085.1 | 2 | 168 | |||
XM_011537784.1 | XP_011536086.1 | 2 | 168 | |||
XM_011537785.1 | XP_011536087.1 | 2 | 168 | |||
XM_011537787.1 | XP_011536089.1 | 2 | 168 | |||
XM_011537788.1 | XP_011536090.1 | 2 | 168 | |||
XM_017018735.1 | XP_016874224.1 | 2 | 168 | |||
XM_017018736.1 | XP_016874225.1 | 2 | 168 | |||
XM_017018737.1 | XP_016874226.1 | 2 | 168 | |||
GenBank mRNA | AY209000.1 | - | 2 | 138 | ||
AY257469.1 | - | 2 | 138 | |||
AY681966.1 | - | 2 | 141 |