Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001729.3 | NP_001720.1 | 4 | 661 | ||
XM_011532211.1 | XP_011530513.1 | 4 | 642 | |||
GenBank mRNA | BC011618.2 | - | 4 | 452 | 3952444 | |
KU177946.1 | - | 4 | 300 | |||
S55606.1 | - | 4 | 594 |