Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZAP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079.3 | NP_001070.2 | 9 | 1193 | ||
NM_207519.1 | NP_997402.1 | 1 | 211 | |||
XM_017004867.1 | XP_016860356.1 | 11 | 2594 | |||
XM_017004868.1 | XP_016860357.1 | 11 | 2576 | |||
XM_017004869.1 | XP_016860358.1 | 11 | 2594 | |||
XM_017004870.1 | XP_016860359.1 | 11 | 2594 | |||
GenBank mRNA | AB083211.1 | - | 1 | 211 | ||
AL832287.1 | - | 3 | 1501 | |||
BC039039.1 | - | 8 | 847 | 5168073 | ||
BC053878.1 | - | 9 | 1169 | 5757161 | ||
L05148.1 | - | 9 | 1195 |