Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005862.2 | NP_001005862.1 | 21, 22 | 2702 | ||
NM_001289936.1 | NP_001276865.1 | 22, 23 | 2753 | |||
NM_001289937.1 | NP_001276866.1 | 18, 19 | 2477 | |||
NM_004448.3 | NP_004439.2 | 18, 19 | 2477 | |||
NR_110535.1 | 18, 19 | 2540 | ||||
GenBank mRNA | AK131568.1 | - | 14, 15 | 3350 | ||
AK295195.1 | - | 22, 23 | 2753 | |||
AK300321.1 | - | 12, 13 | 1651 | |||
AK313683.1 | - | 18, 19 | 2431 | |||
BC080193.1 | - | 18, 19 | 2438 | |||
BC167147.1 | - | 18, 19 | 2219 | 100013444 | ||
HM437238.1 | - | 1, 2 | 66 | |||
KJ534831.1 | - | 18, 19 | 2216 | |||
KJ534964.1 | - | 17, 18 | 2168 | |||
M11730.1 | - | 18, 19 | 2366 | |||
X03363.1 | - | 18, 19 | 2390 |