Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174922.4 | NP_777582.4 | 5 | 441 | ||
XM_006716527.2 | XP_006716590.1 | 5 | 431 | |||
XM_011516907.2 | XP_011515209.1 | 5 | 1154 | |||
XM_011516909.1 | XP_011515211.1 | 5 | 441 | |||
XM_011516910.1 | XP_011515212.1 | 6 | 464 | |||
XM_017013174.1 | XP_016868663.1 | 5 | 479 | |||
GenBank mRNA | AK092773.1 | - | 5 | 404 | ||
BC101659.1 | - | 5 | 381 | 8069165 | ||
BC143326.1 | - | 381 |