Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001348.2 | NP_001339.1 | 3 | 704 | ||
XM_005259508.1 | XP_005259565.1 | 3 | 579 | |||
GenBank mRNA | AB007144.1 | - | 2 | 500 | ||
AB022341.1 | - | 2 | 452 | |||
AK027590.1 | - | 3 | 621 | |||
AK074799.1 | - | 2 | 476 | |||
AK097643.1 | - | 3 | 515 | |||
BC126430.1 | - | 2 | 443 | 8992146 | ||
BC126432.1 | - | 2 | 441 | 8992148 |