Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004073.3 | NP_004064.2 | 2, 3 | 565 | ||
GenBank mRNA | AB209875.1 | - | 3, 4 | 252 | ||
AJ293866.1 | - | 2, 3 | 565 | |||
BC013899.2 | - | 2, 3 | 426 | 4098112 | ||
U56998.1 | - | 2, 3 | 245 |