Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006255.4 | NP_006246.2 | 8 | 1660 | ||
XM_011536954.2 | XP_011535256.1 | 8 | 843 | |||
XM_011536955.1 | XP_011535257.1 | 8 | 776 | |||
XM_011536956.1 | XP_011535258.1 | 8 | 1542 | |||
XM_011536957.1 | XP_011535259.1 | 8 | 1542 | |||
XM_017021458.1 | XP_016876947.1 | 8 | 994 | |||
XM_017021459.1 | XP_016876948.1 | 8 | 1542 | |||
GenBank mRNA | AB451313.1 | - | 8 | 1001 | ||
AK290183.1 | - | 8 | 1379 | |||
AK296158.1 | - | 7 | 674 | |||
AX775931.1 | - | 8 | 736 | |||
AX775933.1 | - | 8 | 1389 | |||
BC037268.1 | - | 8 | 1389 | 4822159 | ||
M55284.1 | - | 8 | 1164 |