Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBXA2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001060.5 | NP_001051.1 | 2 | 643 | ||
NM_201636.2 | NP_963998.2 | 2 | 643 | |||
XM_011528214.2 | XP_011526516.1 | 3 | 2677 | |||
GenBank mRNA | AY429110.1 | - | 1 | 256 | ||
BC028027.1 | - | 2 | 643 | |||
BC074749.2 | - | 1 | 295 | 30915459 | ||
BC074750.2 | - | 1 | 295 | 30915209 | ||
BC121148.1 | - | 1 | 290 | 40121265 | ||
BC121149.2 | - | 291 | ||||
U11271.1 | - | 2 | 468 | |||
U27325.1 | - | 2 | 379 |
Set Membership: |
G-protein coupled receptor |