Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001710.5 | NP_001701.2 | 6 | 1148 | ||
GenBank mRNA | AF349679.1 | - | 6 | 870 | ||
AK130533.1 | - | 6 | 1077 | |||
AK223400.1 | - | 6 | 996 | |||
BC004143.2 | - | 6 | 1005 | 2959705 | ||
BC007990.2 | - | 6 | 1005 | 2959706 | ||
K01566.1 | - | 5 | 609 | |||
L15702.1 | - | 6 | 910 | |||
S67310.1 | - | 6 | 981 | |||
X72875.1 | - | 6 | 870 |