Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAST SIRNA Assays ›
Gene Symbol:
CAST
Gene Name:
calpastatin
Gene Aliases:
BS-17, PLACK
Chromosome Location:
Chr.5: 96662037 - 96774683 on Build GRCh38
UniGene ID:
Hs.436186
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
10389
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAST
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042440.3
NP_001035905.1
18, 19
1678
NM_001190442.1
NP_001177371.1
19, 20
1550
NM_001284212.2
NP_001271141.1
16, 17
1329
NM_001284213.2
NP_001271142.1
15, 16
1236
NM_173060.3
NP_775083.1
17, 18
1386
NR_104285.1
5, 6
542
XM_006714696.3
XP_006714759.1
21, 22
1692
XM_006714697.3
XP_006714760.1
20, 21
1709
XM_006714698.3
XP_006714761.1
19, 20
1670
XM_006714699.3
XP_006714762.1
20, 21
1633
XM_006714700.3
XP_006714763.1
19, 20
1652
XM_006714701.3
XP_006714764.1
19, 20
1668
XM_006714702.3
XP_006714765.1
19, 20
1544
XM_006714703.3
XP_006714766.1
18, 19
1629
XM_006714704.3
XP_006714767.1
19, 20
1577
XM_006714705.3
XP_006714768.1
18, 19
1586
XM_006714706.3
XP_006714769.1
18, 19
1504
XM_006714707.3
XP_006714770.1
18, 19
1455
XM_006714708.3
XP_006714771.1
17, 18
1415
XM_006714709.3
XP_006714772.1
17, 18
1378
XM_006714710.3
XP_006714773.1
16, 17
1333
XM_006714711.3
XP_006714774.1
16, 17
1378
XM_006714712.3
XP_006714775.1
15, 16
1325
XM_006714713.3
XP_006714776.1
5, 6
542
XM_006714714.3
XP_006714777.1
5, 6
542
XM_006714715.3
XP_006714778.1
6, 7
1268
XM_011543654.2
XP_011541956.1
19, 20
1628
XM_011543655.2
XP_011541957.1
19, 20
1625
XM_011543656.2
XP_011541958.1
18, 19
1572
XM_011543657.2
XP_011541959.1
18, 19
1454
XM_011543658.2
XP_011541960.1
16, 17
1307
XM_017009911.1
XP_016865400.1
19, 20
1670
XM_017009912.1
XP_016865401.1
20, 21
1681
XM_017009913.1
XP_016865402.1
20, 21
1650
XM_017009914.1
XP_016865403.1
19, 20
1652
XM_017009915.1
XP_016865404.1
20, 21
1654
XM_017009916.1
XP_016865405.1
19, 20
1668
XM_017009917.1
XP_016865406.1
19, 20
1586
XM_017009918.1
XP_016865407.1
18, 19
1613
XM_017009919.1
XP_016865408.1
18, 19
1586
XM_017009920.1
XP_016865409.1
18, 19
1496
XM_017009921.1
XP_016865410.1
18, 19
1513
XM_017009922.1
XP_016865411.1
17, 18
1547
XM_017009923.1
XP_016865412.1
17, 18
1547
XM_017009924.1
XP_016865413.1
17, 18
1529
XM_017009925.1
XP_016865414.1
17, 18
1465
XM_017009926.1
XP_016865415.1
17, 18
1465
XM_017009927.1
XP_016865416.1
18, 19
1454
XM_017009928.1
XP_016865417.1
17, 18
1453
XM_017009929.1
XP_016865418.1
17, 18
1372
XM_017009930.1
XP_016865419.1
16, 17
1339
XM_017009931.1
XP_016865420.1
16, 17
1333
XM_017009932.1
XP_016865421.1
15, 16
1396
GenBank mRNA
AB208816.1
-
19, 20
1514
AF327443.1
-
14, 15
1228
AK292289.1
-
5, 6
542
AK292356.1
-
5, 6
515
AK296919.1
-
19, 20
1508
AK297879.1
-
17, 18
1328
AK298541.1
-
18, 19
1455
AK299402.1
-
18, 19
1435
AK300624.1
-
15, 16
1219
AK302206.1
-
6, 7
542
AK303836.1
-
11, 12
909
AK311585.1
-
6, 7
720
AK311598.1
-
6, 7
596
AK314290.1
-
16, 17
1331
AK315146.1
-
16, 17
1330
AK315673.1
-
5, 6
490
AL832174.1
-
17, 18
1371
AL832349.1
-
17, 18
1374
AY189741.1
-
7, 8
541
BC013579.1
-
18, 19
1381
3878564
BC078149.1
-
13, 14
964
BT009783.1
-
15, 16
1163
BX648397.1
-
16, 17
1280
D16217.1
-
18, 19
1448
DB033043.1
-
5, 6
490
U26724.2
-
9, 10
811
U31345.1
-
7, 8
558
U38525.1
-
1
14
Back To Top
More Information
Panther Classification:
Molecular Function -
protein-binding activity modulator
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
liver development
egg activation
brain development
myoblast fusion
aging
negative regulation of endopeptidase activity
organ regeneration
myoblast differentiation
negative regulation of cell cycle arrest
cell-cell adhesion
negative regulation of type B pancreatic cell apoptotic process
protease binding
endopeptidase inhibitor activity
protein binding
calcium-dependent cysteine-type endopeptidase inhibitor activity
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-cs4xr:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0