Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003291.2 | NP_003282.2 | 17 | 2160 | ||
XM_005254070.1 | XP_005254127.1 | 17 | 2159 | |||
XM_005254071.1 | XP_005254128.1 | 17 | 2159 | |||
XM_005254072.1 | XP_005254129.1 | 17 | 2159 | |||
GenBank mRNA | AK304313.1 | - | 17 | 2068 | ||
BC039905.1 | - | 18 | 2103 | 6067427 | ||
M55169.1 | - | 17 | 1892 | |||
M73047.1 | - | 17 | 2080 |