Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256134.1 | NP_001243063.1 | 9 | 1250 | ||
NM_003891.2 | NP_003882.1 | 8 | 1184 | |||
XM_017020812.1 | XP_016876301.1 | 10 | 1381 | |||
XM_017020813.1 | XP_016876302.1 | 10 | 1174 | |||
GenBank mRNA | BC074906.2 | - | 8 | 1180 | 30915254 | |
BC074907.2 | - | 8 | 1180 | 30915483 | ||
M55670.1 | - | 8 | 1184 | |||
M55671.1 | - | 9 | 1250 | |||
M59303.1 | - | 6 | 939 |