Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004716.3 | NP_004707.2 | 9 | 1710 | ||
XM_006718940.3 | XP_006719003.1 | 2 | 469 | |||
XM_017018546.1 | XP_016874035.1 | 4 | 238 | |||
GenBank mRNA | AK097207.1 | - | 1 | 885 | ||
AK222907.1 | - | 9 | 1250 | |||
AK312429.1 | - | 9 | 1285 | |||
BC006357.2 | - | 9 | 1228 | 4125594 | ||
BC010696.1 | - | 9 | 1280 | 3868257 | ||
BT006870.1 | - | 7 | 1079 | |||
U33849.1 | - | 9 | 1230 | |||
U40623.1 | - | 7 | 1100 |