Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276479.1 | NP_001263408.1 | 13, 14 | 1745 | ||
NM_001276480.1 | NP_001263409.1 | 12, 13 | 1709 | |||
NM_004793.3 | NP_004784.2 | 12, 13 | 2030 | |||
NR_076392.1 | 13, 14 | 1728 | ||||
GenBank mRNA | AK056366.1 | - | 12, 13 | 1709 | ||
AK096433.1 | - | 13, 14 | 1758 | |||
AK127867.1 | - | 13, 14 | 1810 | |||
AK298531.1 | - | 13, 14 | 1709 | |||
AK309720.1 | - | 13, 14 | 1918 | |||
AL096720.1 | - | 10, 11 | 1642 | |||
BC000235.1 | - | 12, 13 | 1909 | 3350958 | ||
BC109218.1 | - | 12, 13 | 1572 | |||
U02389.1 | - | 12, 13 | 2006 |