Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319214.1 | NP_001306143.1 | 4 | 477 | ||
NM_005040.3 | NP_005031.1 | 5 | 839 | |||
NM_199418.3 | NP_955450.2 | 6 | 902 | |||
XM_005274093.1 | XP_005274150.1 | 5 | 569 | |||
GenBank mRNA | AB451270.1 | - | 5 | 638 | ||
AB451397.1 | - | 5 | 638 | |||
AK091786.1 | - | 6 | 729 | |||
AK302386.1 | - | 4 | 428 | |||
AK312919.1 | - | 5 | 660 | |||
BC001500.2 | - | 5 | 657 | 3510081 | ||
L13977.1 | - | 5 | 667 |