Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319214.1 | NP_001306143.1 | 4 | 563 | ||
NM_005040.3 | NP_005031.1 | 5 | 925 | |||
NM_199418.3 | NP_955450.2 | 6 | 988 | |||
XM_005274093.1 | XP_005274150.1 | 5 | 655 | |||
GenBank mRNA | AK091786.1 | - | 6 | 815 | ||
AK302386.1 | - | 4 | 514 | |||
AK312919.1 | - | 5 | 746 | |||
BC001500.2 | - | 5 | 743 | 3510081 | ||
L13977.1 | - | 5 | 753 |