Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278585.1 | NP_001265514.1 | 19 | 2691 | ||
NM_006587.3 | NP_006578.2 | 21 | 3003 | |||
GenBank mRNA | AF113248.1 | - | 5 | 648 | ||
AF133845.1 | - | 21 | 2939 | |||
AK302816.1 | - | 20 | 2678 | |||
AK304038.1 | - | 20 | 2768 | |||
BC110451.2 | - | 21 | 2939 | 40027293 |