Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001991.2 | NP_001001991.1 | 8, 9 | 1554 | ||
NM_001282675.1 | NP_001269604.1 | 10, 11 | 1544 | |||
NM_001282676.1 | NP_001269605.1 | 6, 7 | 1221 | |||
NM_015430.3 | NP_056245.2 | 9, 10 | 1605 | |||
GenBank mRNA | AF370388.1 | - | 7, 8 | 1043 | ||
AK027841.1 | - | 9, 10 | 1205 | |||
AK297092.1 | - | 10, 11 | 1545 | |||
AK300185.1 | - | 6, 7 | 806 | |||
AK316188.1 | - | 6, 7 | 793 | |||
AL832391.1 | - | 8, 9 | 1120 | |||
AY358346.1 | - | 8, 9 | 1133 | |||
BC089434.1 | - | 9, 10 | 1197 | 30558617 | ||
BX640676.1 | - | 8, 9 | 1554 |