Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPVL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019029.2 | NP_061902.2 | 12 | 1434 | ||
NM_031311.3 | NP_112601.3 | 12 | 1346 | |||
XM_011515437.1 | XP_011513739.1 | 12 | 1513 | |||
XM_017012366.1 | XP_016867855.1 | 10 | 1073 | |||
GenBank mRNA | AF106704.2 | - | 12 | 1266 | ||
AF217508.1 | - | 11 | 1234 | |||
AF282617.1 | - | 12 | 1296 | |||
AK075433.1 | - | 12 | 1347 | |||
AK124472.1 | - | 16 | 1874 | |||
AK225545.1 | - | 12 | 1297 | |||
AY358549.1 | - | 12 | 1323 | |||
BC016838.2 | - | 12 | 1394 | 3888647 |