Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114387.1 | NP_001107859.1 | 5 | 563 | ||
NM_182606.3 | NP_872412.3 | 5 | 572 | |||
GenBank mRNA | AF071882.1 | - | 5 | 523 | ||
AK131518.1 | - | 5 | 572 | |||
AK292950.1 | - | 5 | 501 | |||
AY498712.1 | - | 5 | 475 | |||
BC111796.1 | - | 5 | 489 | 40026103 |