Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000419.4 | NP_000410.2 | 26, 27 | 2848 | ||
XM_011524749.1 | XP_011523051.1 | 26, 27 | 2752 | |||
XM_011524750.1 | XP_011523052.1 | 26, 27 | 2752 | |||
GenBank mRNA | AK315335.1 | - | 26, 27 | 2752 | ||
BC117443.1 | - | 26, 27 | 2746 | 40125994 | ||
BC126442.1 | - | 26, 27 | 2746 | 8992158 | ||
J02764.1 | - | 26, 27 | 2721 | |||
M18085.1 | - | 1, 2 | 121 | |||
M34480.1 | - | 26, 27 | 2752 | |||
X06831.1 | - | 15, 16 | 1547 |