Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000573.3 | NP_000564.2 | 2, 3 | 441 | ||
NM_000651.4 | NP_000642.3 | 2, 3 | 441 | |||
XM_006711166.2 | XP_006711229.1 | 2, 3 | 417 | |||
XM_011509205.1 | XP_011507507.1 | 2, 3 | 417 | |||
GenBank mRNA | AK298486.1 | - | 2, 3 | 332 | ||
AK309342.1 | - | 2, 3 | 422 | |||
BC032550.1 | - | 2, 3 | 330 | |||
X14362.1 | - | 2, 3 | 228 | |||
Y00816.1 | - | 328 |