Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278113.1 | NP_001265042.1 | 11, 12 | 1267 | ||
NM_001278114.1 | NP_001265043.1 | 12, 13 | 1324 | |||
NM_001464.4 | NP_001455.3 | 12, 13 | 1324 | |||
XM_005273468.1 | XP_005273525.1 | 12, 13 | 1262 | |||
XM_011544479.1 | XP_011542781.1 | 8, 9 | 884 | |||
XM_011544480.1 | XP_011542782.1 | 8, 9 | 884 | |||
GenBank mRNA | AJ133005.1 | - | 12, 13 | 1224 | ||
AK301734.1 | - | 12, 13 | 1262 | |||
BC034957.2 | - | 11, 12 | 1187 | 4826530 | ||
BC064547.1 | - | 8, 9 | 867 | 5744846 | ||
U38805.1 | - | 12, 13 | 1288 | |||
U52370.1 | - | 12, 13 | 1227 |