Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001869.2 | NP_001860.2 | 6 | 526 | ||
GenBank mRNA | AK293378.1 | - | 6 | 522 | ||
BC007009.1 | - | 6 | 526 | 3949984 | ||
BC014571.1 | - | 6 | 528 | 3950117 | ||
BC015140.1 | - | 6 | 528 | 4040975 | ||
BT007403.1 | - | 6 | 501 | |||
CD613167.1 | - | 5 | 436 | |||
KU177990.1 | - | 6 | 501 | |||
KU177991.1 | - | 7 | 607 | |||
KU177992.1 | - | 5 | 399 | |||
U19977.1 | - | 6 | 505 |