Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001977.3 | NP_001968.3 | 17 | 2698 | ||
XM_017007877.1 | XP_016863366.1 | 14 | 1376 | |||
GenBank mRNA | AK313412.1 | - | 18 | 2360 | ||
AM888273.1 | - | 17 | 2698 | |||
AM888274.1 | - | 17 | 2698 | |||
AM888275.1 | - | 17 | 2698 | |||
AM888276.1 | - | 17 | 2698 | |||
AM888277.1 | - | 17 | 2698 | |||
AM888278.1 | - | 17 | 2698 | |||
AM888279.1 | - | 17 | 2698 | |||
AM888280.1 | - | 17 | 2698 | |||
AM888281.1 | - | 17 | 2698 | |||
AM888282.1 | - | 17 | 2698 | |||
AM888283.1 | - | 17 | 2698 | |||
BC094770.1 | - | 18 | 2679 | 30528477 | ||
BX641012.1 | - | 14 | 1865 | |||
L12468.1 | - | 18 | 2602 | |||
L14721.1 | - | 18 | 2439 |