Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145938.1 | NP_001139410.1 | 3 | 452 | ||
NM_002421.3 | NP_002412.1 | 3 | 604 | |||
GenBank mRNA | AK024818.1 | - | 3 | 527 | ||
AK098450.1 | - | 4 | 459 | |||
AK130687.1 | - | 3 | 527 | |||
AK223034.1 | - | 2 | 230 | |||
AK223035.1 | - | 3 | 485 | |||
AK223036.1 | - | 1 | 68 | |||
AK223056.1 | - | 2 | 270 | |||
AK223281.1 | - | 3 | 527 | |||
AK225495.1 | - | 2 | 295 | |||
AK297723.1 | - | 3 | 375 | |||
BC013875.2 | - | 3 | 503 | 3834572 | ||
BT006874.1 | - | 3 | 461 | |||
BT020147.1 | - | 3 | 461 | |||
M13509.1 | - | 3 | 529 | |||
X05231.1 | - | 3 | 527 | |||
X54925.1 | - | 3 | 529 |