Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003399.5 | NP_003390.4 | 5, 6 | 651 | ||
GenBank mRNA | AK300954.1 | - | 5, 6 | 672 | ||
BC126174.1 | - | 5, 6 | 568 | 8991890 | ||
BX386512.2 | - | 5, 6 | 650 | |||
U90724.2 | - | 5, 6 | 671 |